Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt1 Rat anti-Human, Clone: DNM-2C1, MilliporeSigma™

Catalog No. MABE306MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE306MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE306MI Supplier MilliporeSigma Supplier No. MABE306
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Dnmt1 Antibody, clone DNM-2C1 is a Rat Monoclonal Antibody for detection of Dnmt1 also known as DNA (cytosine-5)-methyltransferase 1 and has been validated in immunoprecipitation, Chimmunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dnmt1
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DNM-2C1
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2b in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001124295
Gene Symbols DNMT1, AIM, CXXC9, DNMT
Host Species Rat
Immunogen His-tagged recombinant protein corresponding to human Dnmt1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.