Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tRNA aspartic acid methyltransferase 1, Mouse, Clone: 1E12, Abnova™

Catalog No. 89014094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-094 Supplier Abnova Corporation Supplier No. H00001787M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DNMT2.

This gene encodes a protein responsible for the methylation of aspartic acid transfer RNA. While similar in sequence and structure to DNA cytosine methyltransferases, this gene is distinct and highly conserved in its function among taxa. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFHHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVEEKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYE

Specifications

Antigen tRNA aspartic acid methyltransferase 1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1E12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant DNMT2.
Formulation PBS with no preservative; pH 7.4
Gene TRDMT1
Gene Accession No. BC047733
Gene Alias DNMT2/M.HsaIIP/PuMet/RNMT1
Gene Symbols TRDMT1
Host Species Mouse
Immunogen DNMT2 (AAH47733, 1 a.a. ∼ 391 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1787
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.