Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 072050MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-205-0MI 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-205-0MI Supplier MilliporeSigma Supplier No. 072050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-DNMT3A2 Antibody (Rabbit Polyclonal Antibody) validated in immunocytochemistry, ELISA, western blotting to detect DNMT3A2 Antibody also known as DNA (cytosine-5)-methyltransferase 3 alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT3A2
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.
Gene Symbols DNMT3A2 ; Dnmt3a; EC 2.1.1.37; M.HsaIIIA ; OTTHUMP00000120005; OTTHUMP00000120006
Host Species Rabbit
Immunogen The immunogen was a KLH-conjugated synthetic peptide corresponding to the C-terminal region of human DNMT3A2.
Purification Method Protein A chromatography
Quantity 400 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_715640
Target Species Human
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial to pellet the solution. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.