Learn More
DO-11.10 Clonotypic TCR Mouse anti-Mouse, PE, Clone: KJ1-26, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551772
Description
The KJ1-26 antibody reacts with the DO-11.10 Clonotypic T-cell Receptor (TCR) of the BALB/c-derived DO-11.10 T-cell hybridoma and T lymphocytes from the DO-11.10 transgenic mouse (TgN[DO-11.10]10Loh). The DO-11.10 TCR, an 80-90kDa (non-reduced) or 40-44kDa (reduced) protein, is specific for the chicken OVA(323-339)/I-A[d] complex. The DO-11.10 T-cell hybridoma also responds strongly to chicken OVA/I-A[b] and jungle fowl OVA/I-A[d] and weakly to turkey OVA/I-A[d] and I-A[b]. The DO-11.10 mouse model is valuable for studies of T-cell immigration, immunoregulation, development, activation, and function. The KJ1-26 mAb was shown to block the antigen responses of the DO-11.10 T-cell hybridoma in vitro.
Host Species: Mouse
Clone: KJ1-26
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: DO-11.10 T hybridoma cells
Flow Cytometry

Specifications
DO-11.10 Clonotypic TCR | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
DO-11.10 T hybridoma cells | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
KJ1-26 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.