Learn More
Dopa Decarboxylase Rabbit anti-Bovine, Canine, Guinea Pig, Human, Rat, Sheep, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dopa Decarboxylase |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 10mM HEPES, pH 7.5, 150mM NaCl, 100μg/mL BSA and 50% glycerol. |
| Gene Symbols | DDC; AADC; EC 4.1.1.28 |
| Host Species | Rabbit |
| Immunogen | Recombinant bovine DDC expressed in E. coli and purified from inclusion bodies. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.