Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine Mouse anti-Rat, Clone: K56A, MilliporeSigma™

Catalog No. MAB5300MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5300MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5300MI Supplier MilliporeSigma Supplier No. MAB5300

Mouse Monoclonal Antibody

Anti-Dopamine Antibody, clone K56A detects level of Dopamine and has been published and validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine
Applications Immunohistochemistry
Classification Monoclonal
Clone K56A
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing 50% glycerol.
Host Species Mouse
Immunogen Dopamine-Gluteraldehyde-BSA.
Quantity 100 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C for up to 6 months in undiluted aliquots.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.