Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D(1A) Receptor Rabbit anti-Rat, Bovine, Polyclonal, MilliporeSigma™

Catalog No. ABN20MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN20MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN20MI Supplier MilliporeSigma Supplier No. ABN20
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dopamine D(1A) Receptor Antibody detects level of Dopamine D(1A) Receptor and has been published and validated for use in immunohistochemistry, western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine D(1A) Receptor
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols D1a; Drd1; Drd-1; Drd1a
Host Species Rabbit
Immunogen KLH-conjugated linear peptide of rat dopamine D(1A) receptor at and around the cytoplasmic domain.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_036678
Target Species Bovine, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.