Learn More
Description
Specifications
Specifications
| Antigen | Dopamine D1A Receptor |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS with 0.2% BSA and 0.05% sodium azide. |
| Gene Symbols | DRD1; DRD1A; DADR; D |
| Host Species | Rabbit |
| Immunogen | A nine amino acid peptide corresponding to the 4th extracellular domain of the rat D1A receptor. The immunogen is absolutely conserved in the mouse D1A receptor and shows 88% homology with human and 77% with porcine and bovine D1A receptors. The peptide s |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.