Learn More
Dopamine D1A Receptor Rabbit anti-Pig, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dopamine D1A Receptor |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 100mM Tris, pH 7.5 with 0.2% BSA and 0.05% sodium azide. |
| Gene Symbols | DRD1; DRD1A; DADR; D |
| Host Species | Rabbit |
| Immunogen | A 13 amino acid peptide sequence (amino acids 403-415) from rat D1A (Monsma et al. 1990; Zhou et al. 1990; Zhou et al. 1992; O'Dowd et al. 1990). This peptide is predicted to be cytoplasmic. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.