Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D2 Receptor Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1558MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1558MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1558MI Supplier MilliporeSigma Supplier No. AB1558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dopamine D2 Receptor Antibody is an antibody against Dopamine D2 Receptor for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine D2 Receptor
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiSerum. Liquid containing 0.05% sodium azide.
Gene Symbols DRD2; D; D2R; D2DR
Host Species Rabbit
Immunogen Amino-terminal peptide sequence (aa 24-34) near or at the ligand binding domain of rat D2R (1,2). The immunogen shows 100% homology with mouse 90% with human and bovine and 72% with canine D2R receptors. An antibody made to the same sequence has shown
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000795.2; NM_016574.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.