Learn More
Description
Specifically detects Dopamine D2 Receptor Clone: in Human, Rat samples, and it is validated for ELISA, Immunohistochemistry, Immuno Blotting
Specifications
Specifications
| Antigen | Dopamine D2 Receptor |
| Applications | ELISA, Immunoblot, Immunoblot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Undiluted serum. |
| Host Species | Rabbit |
| Immunogen | a synthetic peptide (CAARRAQELEME) corresponding to amino acids 272-282 of human D2 dopamine receptor |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.