Learn More
Description
Specifically detects Dopamine D3 Receptor Clone: in Human, Rat samples, and it is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immuno Blotting
Specifications
Specifications
| Antigen | Dopamine D3 Receptor |
| Applications | ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Undiluted serum. |
| Host Species | Rabbit |
| Immunogen | a synthetic peptide (ASLSQLSSHC) corresponding to amino acids (2-10) of human dopamine D3 receptor |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.