Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D4 Receptor Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB1787PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1787PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1787PMI Supplier MilliporeSigma Supplier No. AB1787P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dopamine D4 Receptor Antibody, extracellular domain is an antibody against Dopamine D4 Receptor for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine D4 Receptor
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA
Gene Symbols DRD4; D4DR; D
Host Species Rabbit
Immunogen A 10 amino acid peptide sequence from the mouse D4 receptor within the 3rd extracellular domain.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000797.2
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.