Learn More
Dopamine Β Hydroxylase Rabbit anti-Bovine, Guinea Pig, Human, Rat, Rabbit, Feline, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dopamine β Hydroxylase |
| Applications | Immunohistochemistry, Radioimmune Assays (RIA), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit Serum. Lyophilized, no preservatives. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any insoluble material. |
| Gene Symbols | DBH; DBM; EC 1.14.17.1 [Contains: Soluble dopamine beta-hydroxylase]. |
| Host Species | Rabbit |
| Immunogen | Native enzyme from bovine adrenal glands. |
| Purification Method | Unpurified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.