Learn More
Dopamine Receptor D1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dopamine Receptor D1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide. |
| Gene Symbols | DRD1; D; DRD1A; DADR |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the C-terminal domain of human Dopamine Receptor D1. The immunogen has 94% conservation with rat and 88% with mouse. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.