Learn More
Dopamine Transporter Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dopamine Transporter |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified immunoglobulin. Liquid in PBS containing 0.1% BSA |
| Gene Symbols | SLC6A3; ADHD; DAT; DAT1; ADD |
| Host Species | Rabbit |
| Immunogen | An 18 amino acid peptide sequence near the NH2-terminus of rat brain dopamine transporter coupled to KLH (Kilty et al. 1991; Giros et al. 1991; Shimada et al. 1991; Kitayma 1992). This region is presumably the cytoplasmic domain. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.