Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine Transporter Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB1766MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1766MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1766MI Supplier MilliporeSigma Supplier No. AB1766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dopamine Transporter Antibody, C-terminus detects level of Dopamine Transporter and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine Transporter
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 100μL of 10mM HEPES (pH 7.5), 150mM NaCl, 100μg/mL BSA, and 50% glycerol.
Gene Symbols SLC6A3; ADHD; DAT; DAT1; ADD
Host Species Rabbit
Immunogen Synthetic peptide from the intracellular C-terminal region of human dopamine transporter (CEKDRELVDRGEVRQFTLRHWL).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001044.3
Target Species Human, Monkey
Content And Storage −20-−80°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.