Learn More
Dopamine Transporter Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dopamine Transporter |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Each vial contains ∼33 μg of affinity-purified antibody in 100μL of 10mM HEPES (pH 7.5), 150mM NaCl with 100μg/mL BSA and 50% glycerol. |
| Gene Symbols | SLC6A3; ADHD; DAT; DAT1; ADD |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the EL2 region of human dopamine transporter. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.