Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine Transporter; 100 μg Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2231MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2231MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2231MI Supplier MilliporeSigma Supplier No. AB2231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Dopamine Transporter, 100 μg Antibody is validated for use in western blotting for the detection of Dopamine Transporter, 100 μg.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine Transporter
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified Antibody in PBS with 0.1% NaN3
Gene Symbols DAT ; DAT 1
Host Species Rabbit
Immunogen Synthetic Linear Peptide
Purification Method Immunoaffinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001044.3
Target Species Mouse, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Prior to removing the cap gently centrifuge the vial and gently mix the solution.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.