Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine Transporter Rat anti-Human, Mouse, Monkey, Rat, Clone: DAT-Nt, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MAB369
Description
This Anti-Dopamine Transporter Antibody, N-terminus, clone DAT-Nt is validated for use in immunocytochemistry, immunohistochemistry, western blotting for the detection of Dopamine Transporter.
Specifications
Dopamine Transporter | |
Monoclonal | |
Unconjugated | |
NM_001044.3 | |
Rat | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2a κ |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
DAT-Nt | |
UnPurified tissue culture supernatant, filtered through a 0.2μm membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL. | |
SLC6A3, DAT1, ADHD, ADD, DAT | |
N-terminus of human dopamine transporter fused to Glutathione S-transferase. | |
100 μL | |
Human, Monkey, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction