Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doublecortin Guinea Pig anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB2253MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2253MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2253MI Supplier MilliporeSigma Supplier No. AB2253
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

This Anti-Doublecortin Antibody is validated for use in western blot, immunohistochemistry, IC for the detection of Doublecortin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Doublecortin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation GST-Depleted Sera with 0.05% NaN3 .
Gene Symbols DBCN ; DC ; Doublin ; LISX ; Lis-X; Lissencephalin-X; OTTHUMP00000062892; SCLH; XLIS ; doublecortex; doublecortin; doublin ; lissencephalin-X
Host Species Guinea Pig
Immunogen Recombinant Peptide.
Purification Method GST Depletion
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000555.2
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.