Learn More
DP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA129135
Catalog No. PIPA129135
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
PA1-29136 detects E2F4 from human, rat, mouse samples. PA1-29136 has been successfully used in Western blot applications. The PA1-29136 immunogen is: Synthetic peptide from the C terminus of human E2F4 protein.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DP2
Polyclonal
Unconjugated
Q14188, Q64163
TFDP2
Synthetic peptide corresponding to the C-terminus of human DP-2 protein
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
1 mg/mL
TFDP2
BTPS2, DP2, DP2.5, DP3, GS
Rabbit
Protein A
RUO
211586, 300947, 7029
4° C, do not freeze
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.