Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPAGT1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526966
Description
This antibody is predicted to react with non-human primate based on sequence homology.
DPAGT1 encoded by this gene is an enzyme that catalyzes the first step in the dolichol-linked oligosaccharide pathway for glycoprotein biosynthesis. This enzyme belongs to the glycosyltransferase family 4. This protein is an integral membrane protein of the endoplasmic reticulum. The congenital disorder of glycosylation type Ij is caused by mutation in the gene encoding this enzyme.
Specifications
| DPAGT1 | |
| Polyclonal | |
| DPAGT1 | |
| ALG7, CDG-Ij, CDG1J, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT, GlcNAc-1-P transferase, N-acetylglucosamine-1-phosphate transferase, UDP-GlcNAc:dolichyl-phosphate N-acetylglucosaminephosphotransferase, UDP-N-acetylglucosamine--dolichyl-phosphate N-acetylglucosaminephosphotransferase, dolichyl-phosphate (UDP-N-acetylglucos | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1798 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Q9H3H5 | |
| DPAGT1 | |
| KLH conjugated synthetic peptide between 286-315 amino acids from the central region of human DPAGT1 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction