Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPA-2 Mouse anti-Mouse, FITC, Clone: 6C1.2, MilliporeSigma™

Catalog No. FCMAB136FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB136FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB136FMI Supplier MilliporeSigma Supplier No. FCMAB136F

Mouse Monoclonal Antibody

Milli-Mark™ Anti-DPPA-2-FITC Antibody, clone 6C1.2 detects level of DPPA-2 and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA-2
Applications Flow Cytometry
Classification Monoclonal
Clone 6C1.2
Conjugate FITC
Formulation Purified mouse monoclonal IgG2a in buffer containing PBS, 15mg/mL BSA with 0.1% sodium azide.
Gene Accession No. NP_082891
Gene Symbols PESCRG1
Host Species Mouse
Immunogen Full length recombinant mouse DPPA-2 protein
Purification Method Protein G Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.