Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPA4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen DPPA4
Concentration 0.5 mg/mL
Dilution Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224656SS
SDP
View Documents
Novus Biologicals
NBP224656SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224656
SDP
View Documents
Novus Biologicals
NBP224656
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

DPPA4 Polyclonal specifically detects DPPA4 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DPPA4
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Polyclonal
Purified
RUO
Human, Primate
Q7L190
55211
A portion of amino acid 1 to 50 of human DPPA4 was used as immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Stem Cell Markers
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
developmental pluripotency associated 4, developmental pluripotency-associated protein 4, FLJ10713,2410091M23Rik
DPPA4
IgG
Protein A purified
The amino acid sequence used as immunogen for the hnRNPK antibody is 100% homologous in human, mouse, monkey, chimpanzee and rat, 94% homologous in dog and 78% homologous in chicken.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.