Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dihydropyrimidine dehydrogenase, Mouse, Clone: 7D4, Abnova™

Catalog No. 89015826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-015-826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-015-826 Supplier Abnova Corporation Supplier No. H00001806M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant DPYD.

The protein encoded by this gene is a pyrimidine catabolic enzyme and the initial and rate-limiting factor in the pathway of uracil and thymidine catabolism. Mutations in this gene result in dihydropyrimidine dehydrogenase deficiency, an error in pyrimidine metabolism associated with thymine-uraciluria and an increased risk of toxicity in cancer patients receiving 5-fluorouracil chemotherapy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MAPVLSKDSADIESILALNPRTQTHATLCSTSAKKLDKKHWKRNPDKNCFNCEKLENNFDDIKHTTLGERGALREAMRCLKCADAPCQKSCPTNLDIKSFITSIANKNYY

Specifications

Antigen dihydropyrimidine dehydrogenase
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 7D4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant DPYD.
Formulation PBS with no preservative; pH 7.4
Gene DPYD
Gene Accession No. NM_000110
Gene Alias DHP/DHPDHASE/DPD/MGC132008/MGC70799
Gene Symbols DPYD
Host Species Mouse
Immunogen DPYD (NP_000101, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1806
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.