Learn More
DRD5, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DRD5 peptide using ARM Technology.
Supplier: Abnova Corporation H00001816K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes the D5 subtype of the dopamine receptor. The D5 subtype is a G-protein coupled receptor which stimulates adenylyl cyclase. This receptor is expressed in neurons in the limbic regions of the brain. It has a 10-fold higher affinity for dopamine than the D1 subtype. Pseudogenes related to this gene reside on chromosomes 1 and 2. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DRD5 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DRD5 | |
DRD5 | |
A synthetic peptide of human DRD5 | |
RUO | |
1816 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
dopamine receptor D5 | |
DRD5 | |
DBDR, DRD1B, DRD1L2, MGC10601 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.