Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drp1 (Density Regulated Protein-1) Mouse, Unlabeled, Clone: 22, BD

Catalog No. BDB611739 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611739 150 μg
BDB611738 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611739 Supplier BD Biosciences Supplier No. 611739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In culture, cell density can have profound effects on gene expression, enzymatic activity, and cell signaling pathways. Using differential screening of cDNAs from low-passage nontumorigenic teratocarcinoma cells versus high passage tumorigenic cells, a protein was identified that is regulated in cell-density dependent manner. This protein, density-regulated protein 1 (drp1), contains putative sites for N-myristoylation and phosphorylation sites for cAMP and/or cGMP-dependent kinase, casein kinase II, and PKC. The expression of drp1 is enriched in high density cultures of both nontumorigenic and tumorigenic cell lines and is widely detected in adult organs, especially skeletal and cardiac muscle. In addition, increased expression of drp1 is not due to growth arrest as a result of serum starvation or TGF-β treatment nor is it a result of factors found in the media of high density cultures. Interestingly, drp1 is expressed highest in skeletal and cardiac muscle where unique cell-cell contacts are involved in muscle cell membrane depolarization and contraction. Thus, drp1 expression may be regulated by signaling pathways related to specific types of cell-cell contacts.

Host Species: Mouse
Clone: 22
Isotype: IgG2a
Species Reactivity [for Features Main]: Human
Immunogen: Human Drp1 aa. 19-201

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Drp1 (Density Regulated Protein-1)
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Description Density Regulated Protein-1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Drp1 aa. 19-201
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.