Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DsbC, Mouse, Clone: 2C2, Abnova™

Catalog No. 89024818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-818 Supplier Abnova Corporation Supplier No. MAB0045M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant DsbC.

Specifications

Antigen DsbC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant DsbC.
Host Species Mouse
Immunogen DsbC (NP_457441, 22 a.a. ∼ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Unpurified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.