Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCAN family member 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human DSCR1L2 protein.
Supplier: Abnova Corporation H00011123B01
Description
Sequence: MLRDTMKSWNDSQSDLCSTDQEEEEEMIFGENEGDLDEMMDLSDLPTSLFACSVHEAVFEAREQKERFEALFTIYDDQVTFQLFKSFRRVRINFSKPEAAARARIELHETDFNGQKLKLYFAQVQMSGEVRDKSYLLPPQPVKQFLISPPASPPVGWKQSEDAMPVINYDLLCAVSKLGPGEKYELHAGTESTPSVVVHVCESETEEEEETKNPKQKIAQTRRPDPPTAALNEPQTFDCALSpecifications
RCAN family member 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human DSCR1L2 protein. | |
RCAN3 | |
DSCR1L2/MCIP3/RCN3/hRCN3 | |
Mouse | |
50 μL | |
Yes | |
11123 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC035854 | |
RCAN3 | |
DSCR1L2 (AAH35854, 1 a.a. ∼ 241 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction