Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSG2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT181
Description
Detect Desmoglein using this rabbit polyclonal antibody, Anti-DSG2 Antibody validated for use in western blotting and immunoprecipitation.
Specifications
DSG2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunoprecipitation, Western Blot | |
Unconjugated | |
DSG2; CDHF5 | |
KLH-conjugated linear peptide corresponding to human DSG2 near the C-terminus. | |
100 μg | |
NP_001934 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction