Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTNBP1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89003204 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-204 Supplier Abnova Corporation Supplier No. H00084062D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human DTNBP1 protein.

This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations in this gene are associated with Hermansky-Pudlak syndrome type 7. This gene may also be associated with schizophrenia. Multiple transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MLETLRERLLSVQQDFTSGLKTLSDKSREAKVKSKPRTVPFLPKYSAGLELLSRYEDTWAALHRRAKDCASAGELVDSEVVMLSAHWEKKKTSLVELQEQLQQLPALIADLESMTANLTHLEASFEEVENNLLHLEDLCGQCELERCKHMQSQQLENYKKNKRKELETFKAELDAEHAQKVLEMEHTQQMKLKERQKFFEEAFQQDMEQYLSTGYLQIAERREPIGSMSSMEVNVDMLEQMDLMDISDQEALDVFLNSGGEENTVLSPALGPESSTCQNEITLQVPNPSELRAKPPSSSSTCTDSATRDISEGGESPVVQSDEEEVQVDTALATSHTDREATPDGGEDSDS

Specifications

Antigen DTNBP1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human DTNBP1 protein.
Formulation PBS with no preservative; pH 7.4
Gene DTNBP1
Gene Accession No. NM_032122.3
Gene Alias DBND/DKFZp564K192/FLJ30031/HPS7/MGC20210/My031/SDY
Gene Symbols DTNBP1
Host Species Rabbit
Immunogen DTNBP1 (NP_115498.2, 1 a.a. ∼ 351 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84062
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.