Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dual oxidase 2 Mouse anti-Human, Clone: Duox S-12, MilliporeSigma™

Catalog No. MABN787MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN787MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN787MI Supplier MilliporeSigma Supplier No. MABN787
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Dual oxidase 2, clone Duox S-12 Antibody is validated for use in Western Blotting and Immunohistochemistry and Immunocytochemistry for the detection of Dual oxidase 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dual oxidase 2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Duox S-12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_054799
Gene Symbols DUOX2, LNOX2, THOX2
Host Species Mouse
Immunogen Recombinant protein corresponding to the N-terminus of human Dual oxidase 2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.