Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ductin Rabbit anti-Invertebrates, Vertebrates, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5469
Description
Anti-Ductin Antibody detects level of Ductin and has been published and validated for use in immunocytochemistry, immunohistochemistry and western blotting.
Specifications
Ductin | |
Polyclonal | |
Affinity purified. Liquid in 50% glycerol. Contains no preservative. | |
Rabbit | |
Affinity Purified | |
Primary | |
Invertebrate, Vertebrates |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
ATP6V0C; VATL; Vma3; ATPL; ATP6C; ATP6L; EC 3.6.3.14 | |
Synthetic peptide (100% conserved in all species) cooresponding to amino acids 115 to 126 of human ATP6VOC {gb AAH07389.1}. | |
100 μL | |
NM_001694.2 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction