Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dual specificity phosphatase 26 (putative), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008141 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-141 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-141 Supplier Abnova Corporation Supplier No. H00078986B01

Mouse polyclonal antibody raised against a full-length human DUSP26 protein.

Sequence: MCPGNWLWASMTFMARFSRSSSRSPVRTRGTLEEMPTVQHPFLNVFELERLLYTGKTACNHADEVWPGLYLGDQDMANNRRELRRLGITHVLNASHSRWRGTPEAYEGLGIRYLGVEAHDSPAFDMSIHFQTAADFIHRALSQPGGKILVHCAVGVSRSATLVLAYLMLYHHLTLVEAIKKVKDHRGIIPNRGFLRQLLALDRRLRQGLEA

Specifications

Antigen dual specificity phosphatase 26 (putative)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DUSP26 protein.
Formulation No additive
Gene DUSP26
Gene Accession No. NM_024025.1
Gene Alias DUSP24/LDP-4/MGC1136/MGC2627/MKP8/NATA1/SKRP3
Gene Symbols DUSP26
Host Species Mouse
Immunogen DUSP26 (NP_076930.1, 1 a.a. ∼ 211 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 78986
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.