Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dual specificity phosphatase 4, Mouse, Clone: 1D12, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant DUSP4.

Supplier:  Abnova Corporation H00001846M08

Catalog No. 89-000-612


Only null left
Add to Cart

Description

Description

The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, ERK2 and JNK, is expressed in a variety of tissues, and is localized in the nucleus. Two alternatively spliced transcript variants, encoding distinct isoforms, have been observed for this gene. In addition, multiple polyadenylation sites have been reported. [provided by RefSeq

Sequence: MVTMEELREMDCSVLKRLMNRDENGGGAGGSGSHGTLGLPSGGKCLLLDCRPFLAHSAGYILGSVNVRCNTIVRRRAKGSVSLEQILPAEEEVRARLRSGLYSAVIVYDERSPRAESLREDSTVSLVVQALRRNAERTDICLLKGGYERFSSEYPEFCSKTKALAAIPPPVPPSATEPLDLGCSSCGTPLHDQGGPVEILPFLYLGSAYHAARRDMLDALGITALLNVSSDCPNHFEGHYQYKCIPVEDNHKADISSWFMEAIEYIDAVKDCRGRVLVHCQAGISRSATICLAYLMMKKRVRLEEAFEFVKQRRSIISPNFSFMGQLLQFESQVLATSCAAEAASPSGPLRERGKTPATPTSQFVFSFPVSVGVHSAPSSLPYLHSPITTSPSC
Specifications

Specifications

dual specificity phosphatase 4
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
BC002671
DUSP4
DUSP4 (AAH02671, 1 a.a. ∼ 394 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Cell Cycle
Primary
Human
Antibody
ELISA, Immunofluorescence, Western Blot
1D12
Mouse monoclonal antibody raised against a full length recombinant DUSP4.
DUSP4
HVH2/MKP-2/MKP2/TYP
Mouse
Affinity Purified
RUO
Yes
1846
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.