Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

deoxyuridine triphosphatase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017578 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-578 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-578 Supplier Abnova Corporation Supplier No. H00001854D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human DUT protein.

This gene encodes an essential enzyme of nucleotide metabolism. The encoded protein forms a ubiquitous, homotetrameric enzyme that hydrolyzes dUTP to dUMP and pyrophosphate. This reaction serves two cellular purposes: providing a precursor (dUMP) for the synthesis of thymine nucleotides needed for DNA replication, and limiting intracellular pools of dUTP. Elevated levels of dUTP lead to increased incorporation of uracil into DNA, which induces extensive excision repair mediated by uracil glycosylase. This repair process, resulting in the removal and reincorporation of dUTP, is self-defeating and leads to DNA fragmentation and cell death. Alternative splicing of this gene leads to different isoforms that localize to either the mitochondrion or nucleus. A related pseudogene is located on chromosome 19. [provided by RefSeq

Sequence: MTPLCPRPALCYHFLTSLLRSAMQNARGARQRAEAAVLSGPGPPLGRAAQHGIPRPLSSAGRLSQGCRGASTVGAAGWKGELPKAGGSPAPGPETPAISPSKRARPAEVGGMQLRFARLSEHATAPTRGSARAAGYDLYSAYDYTIPPMEKAVVKTDIQIALPSGCYGRVAPRSGLAAKHFIDVGAGVIDEDYRGNVGVVLFNFGKEKFEVKKGDRIAQLICERIFYPEIEEVQALDDTERGSGGFGSTGKN

Specifications

Antigen deoxyuridine triphosphatase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human DUT protein.
Formulation PBS with no preservative; pH 7.4
Gene DUT
Gene Accession No. NM_001025248
Gene Alias FLJ20622/dUTPase
Gene Symbols DUT
Host Species Rabbit
Immunogen DUT (NP_001020419.1, 1 a.a. ∼ 252 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1854
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.