Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dux4 Mouse anti-Human, Clone: 9A12, MilliporeSigma™

Catalog No. MABD116MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD116MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD116MI Supplier MilliporeSigma Supplier No. MABD116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Dux4 using this mouse monoclonal antibody, Anti-Dux4 Antibody, clone 9A12 validated for use in western blotting, immunocytochemistry, Electrophoretic Mobility Shift Assay and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dux4
Applications Electromobility Shift Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9A12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_149418
Gene Symbols DUX4, DUX10
Host Species Mouse
Immunogen recombinant protein corresponding to the domain of: C-Terminal region of Human Dux4.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.