Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin 1 Mouse anti-Human, Mouse, Rat, Clone: D5, MilliporeSigma™

Catalog No. MAB5402MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5402MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5402MI Supplier MilliporeSigma Supplier No. MAB5402

Mouse Monoclonal Antibody

This Anti-Dynamin 1 Antibody, neuronal specifimmunocytochemistry, clone D5 is validated for use in western blot, immunohistochemistry for the detection of Dynamin 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynamin 1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone D5
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing 0.1% sodium azide.
Gene Accession No. NM_001005336.1; NM_004408.2
Gene Symbols DNM1, dynamin, dynamin1, Dynamin-1, DNM, EC 3.6.5.5
Host Species Mouse
Immunogen Recombinant His-tagged fusion protein, corresponding to amino acids 1-750 of Human Dynamin 1.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype Ig
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.