Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynein Mouse anti-Human, Mouse, Rat, Bovine, Drosophila, Sheep, Fish, Xenopus, Clone: 74.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1618
Description
Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmimmunocytochemistry, clone 74.1 detects level of Dynein and has been published and validated for use in immunocytochemistry, immunofluoresence, immunoprecipitation and western blotting.
Specifications
Dynein | |
Monoclonal | |
Unconjugated | |
NM_012144.2 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
74.1 | |
Purified mouse monoclonal IgG2b in buffer containing 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. | |
DNAI1, PCD, ICS, MGC26204, CILD1 | |
Bovine brain cytoplasmic dynein. | |
100 μg | |
Bovine, Drosophila, Fish, Human, Mouse, Rat, Sheep, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction