Learn More
DYRK1A, Mouse, Clone: 7D10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DYRK1A.
Supplier: Abnova Corporation H00001859M01
Description
This gene encodes a member of the Dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) family. This member contains a nuclear targeting signal sequence, a protein kinase domain, a leucine zipper motif, and a highly conservative 13-consecutive-histidine repeat. It catalyzes its autophosphorylation on serine/threonine and tyrosine residues. It may play a significant role in a signaling pathway regulating cell proliferation and may be involved in brain development. This gene is a homolog of Drosophila mnb (minibrain) gene and rat Dyrk gene. It is localized in the Down syndrome critical region of chromosome 21, and is considered to be a strong candidate gene for learning defects associated with Down syndrome. Alternative splicing of this gene generates several transcript variants differing from each other either in the 5' UTR or in the 3' coding region. These variants encode at least five different isoforms. [provided by RefSeq]
Sequence: NQGNQAYQNRPVAANTLDFGQNGAMDVNLTVYSNPRQETGIAGHPTYQFSANTGPAHYMTEGHLTMRQGADREESPMTGVCVQQSPVASSSpecifications
DYRK1A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001396 | |
DYRK1A | |
DYRK1A (NP_001387, 674 a.a. ∼ 763 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
7D10 | |
Mouse monoclonal antibody raised against a partial recombinant DYRK1A. | |
DYRK1A | |
DYRK/DYRK1/HP86/MNB/MNBH | |
Mouse | |
Affinity chromatography | |
RUO | |
1859 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.