Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2, Mouse, Clone: 6E2, Abnova™

Catalog No. 89001080 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-080 Supplier Abnova Corporation Supplier No. H00008445M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant DYRK2.

DYRK2 belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development. The family is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 has demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. Two isoforms of DYRK2 have been isolated. The predominant isoform, isoform 1, lacks a 5' terminal insert. [provided by RefSeq]

Sequence: MNDHLHVGSHAHGQIQVQQLFEDNSNKRTVLTTQPNGLTTVGKTGLPVVPERQLDSIHRRQGSSTSLKSMEGMGKVKATPMTPEQAMKQYMQKLTAFEHH

Specifications

Antigen dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant DYRK2.
Formulation PBS with no preservative; pH 7.4
Gene DYRK2
Gene Accession No. BC005809
Gene Alias FLJ21217/FLJ21365
Gene Symbols DYRK2
Host Species Mouse
Immunogen DYRK2 (AAH05809, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8445
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.