Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrobrevin Mouse, Unlabeled, Clone: 23, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610766
Description
Acetylcholine receptor clustering is considered an early event and a marker for synaptogenesis between a motor neuron and a muscle fiber. This process is mediated by the extracellular protein Agrin which binds to α; and β; Dystroglycan, previously discovered as Dystrobrevin- associated proteins. Dystrobrevin binds to the actin cytoskeleton by its N-terminal domain, thus providing the link between the actin cytoskeleton and receptor clustering. In Torpedo electric organ, Dystrobrevin is a prominent phosphotyrosine-containing protein. Murine Dystrobrevins comprise a family of four related proteins (1-4) with similar function, generated by alternative splicing. Dystrobrevin-1 is 688 amino acids and has an apparent mobility of 87kDa. Dystrobrevin-2 shares almost identical sequence with Dystrobrevin-1 in the first 500 amino acids and has an apparent mobility of 59kDa.
Specifications
Dystrobrevin | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse Dystrobrevin aa. 249-403 | |
50 μg | |
Cell Biology | |
Mouse, Rabbit, Rat | |
IgG1 |
Western Blot | |
23 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction