Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Dystroglycan Mouse anti-Canine, Guinea Pig, Human, Mouse, Rat, Rabbit, Clone: VIA4-1, MilliporeSigma™

Catalog No. 05298MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-298-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-298-MI Supplier MilliporeSigma Supplier No. 05298
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect α-Dystroglycan using this Anti-α-Dystroglycan Antibody, clone VIA4-1. This antibody has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Dystroglycan
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone VIA4-1
Conjugate Unconjugated
Formulation Culture supernatant containing 0.05% sodium azide.
Gene Accession No. NM_004393
Gene Symbols DAG1, A3a, DAG, beta-dystroglycan, alpha-dystroglycan, AGRNR, 156DAG
Host Species Mouse
Immunogen Rabbit skeletal muscle membrane preparation
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Canine, Guinea Pig, Human, Mouse, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.