Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrophin Mouse anti-Canine, Human, Mouse, Rat, Rabbit, Clone: 6D3, MilliporeSigma™

Catalog No. MAB1692MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1692MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1692MI Supplier MilliporeSigma Supplier No. MAB1692
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Dystrophin Antibody, mid-rod, clone 6D3 is validated for use in western blot, immunohistochemistry for the detection of Dystrophin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dystrophin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6D3
Conjugate Unconjugated
Formulation Culture supernatant, liquid in PBS with 1% BSA, containing 15mM sodium azide.
Gene Accession No. NM_000109.2; NM_004006.1; NM_004007.1; NM_004009.2; NM_004010.2; NM_004011.2; NM_004012.2; NM_004013.1; NM_004014.1; NM_004015.1; NM_004016.1; NM_004017.1; NM_004018.1; NM_004019.2; NM_004020.1; NM_004021.1; NM_004022.1; NM_004023.1
Gene Symbols DMD, DXS272, DXS269, DXS230, Dystrophin., DXS268, DXS270, DXS239, DXS142, CMD3B, DXS206, dystrophin, DXS164, BMD
Host Species Mouse
Immunogen Bacterial fusion protein (Cell (1987) 51:919-928).
Quantity 1 mL
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.