Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 610405

 View more versions of this product

Catalog No. BDB610405


Description

Description

E-Cadherin is a 120kDa transmembrane glycoprotein that is localized in the adherens junctions of epithelial cells. There, it interacts with the cytoskeleton through the associated cytoplasmic catenin proteins. In addition to being a calcium-dependent adhesion molecule, E-Cadherin is a critical regulator of epithelial junction formation. Its association with catenins is necessary for cell-cell adhesion. These E-cadherin/catenin complexes associate with cortical actin bundles at both the zonula adherens and the lateral adhesion plaques. Tyrosine phosphorylation can disrupt these complexes, leading to changes in cell adhesion properties. E-Cadherin expression is often down-regulated in highly invasive, poorly differentiated carcinomas. Increased expression of E-Cadherin in these cells reduces invasiveness. Thus, loss of expression or function of E-Cadherin appears to be an important step in tumorigenic progression.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

E-Cadherin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Human, Rat
IgG2b
Western Blot
34
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.