Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Mouse anti-Human, Clone: 67A4, MilliporeSigma™

Catalog No. MAB3199MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3199MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3199MI Supplier MilliporeSigma Supplier No. MAB3199
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-E-Cadherin Antibody, clone 67A4 detects level of E-Cadherin and has been published and validated for use in FC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E-Cadherin
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 67A4
Conjugate Unconjugated
Formulation Protein A purified immunoglobulin. Liquid in 0.02M PBS with NaCl, pH=7.6, 0.1% Sodium azide.
Gene Accession No. NM_004360.2
Gene Symbols CDH1, CDHE, E-cadherin, Arc-1, Uvomorulin, UVO, Cadherin-1, CD324, uvomorulin, ECAD, LCAM
Host Species Mouse
Immunogen T-47D breast carcinoma line
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.