Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Mouse anti-Human, Clone: 67A4, MilliporeSigma™

Catalog No. MAB3199ZCMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3199ZCMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3199ZCMI Supplier MilliporeSigma Supplier No. MAB3199ZC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-E-Cadherin Antibody, clone 67A4, Azide and Ascites Free is validated for use in western blotting, flow cytometry, inhibition and immunofluorescence for the detection of E-Cadherin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E-Cadherin
Applications Flow Cytometry, Immunofluorescence, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 67A4
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.
Gene Accession No. NP_004351
Gene Symbols CDH1, CDHE, UVO
Host Species Mouse
Immunogen Whole cells corresponding to human E-Cadherin.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.