Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-cadherin Rat anti-Human, Mouse, Clone: Decma-1, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT26
Description
This Anti-E-cadherin Antibody, clone DECMA-1 is validated for use in western blot, immunohistochemistry, immunocytochemistry, immunoprecipitation, flow cytometry, FUNC for the detection of E-cadherin.
Specifications
E-Cadherin | |
Monoclonal | |
Unconjugated | |
NP_033994 | |
Rat | |
Protein G | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Decma-1 | |
Purified rat monoclonal IgG1κ in buffer containing PBS without preservative. | |
Arc-1, CD324, CDHE, Cadherin-1, E-cadherin, ECAD, LCAM, UVO, Uvomorulin, uvomorulin | |
Mouse embryonal carcinoma cell line PCC4 | |
100 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction