Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli serotype K88Ab Pili Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA185411
Description
Sheep anti Escherichia coli K88AB pili antibody recognizes the AB variant og the K88 pilus antigen (Mooi & De Graaf 1979).
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications
E. coli serotype K88Ab Pili | |
Polyclonal | |
Unconjugated | |
Escherichia coli | |
E. coli strain 08:K87:K88ab was used. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Sheep | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction